missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dioxolane, 99.5+%, pure, stabilized

Product Code. p-3736365 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
Unit Size:
1 L
2.5 L
25 L
This item is not returnable. View return policy

Product Code. 10227560

Brand: Thermo Scientific Acros 299480010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 646-06-0 | C3H6O2 | 74.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 646-06-0, 128-37-0
Molecular Formula C3H6O2
Molecular Weight (g/mol) 74.08
MDL Number MFCD00003207
InChI Key WNXJIVFYUVYPPR-UHFFFAOYSA-N
Synonym dioxolane, glycolformal, formal glycol, 1,3-dioxolan, 1,3-dioxacyclopentane, 1,3-dioxalane, ethylene glycol formal, glycol methylene ether, glycol formal, dioxolan czech
PubChem CID 31404
ChEBI CHEBI:34247
IUPAC Name 2,6-ditert-butyl-4-methylphenol
SMILES CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C

Specifications

CAS Min % 99.4
CAS Max % 100.0
Melting Point -95°C
Color White
Density 1.0600g/mL
Boiling Point 74.0°C to 75.0°C
Flash Point −6°C
Infrared Spectrum Authentic
Assay Percent Range 99.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.3990 to 1.4010
Quantity 1 L
Beilstein 19, 2
Fieser 14,281
Specific Gravity 1.06
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,ether and acetone
Stabilizer 75ppm BHT max.
Viscosity 0.66 mPa.s (20°C)
Formula Weight 74.08
Percent Purity 99.5+%
Grade Pure
Physical Form Crystalline Powder
Chemical Name or Material 1, 3-Dioxolane, Stabilized
Show More Show Less
compliance-icons
Product Identifier
  • 1,3-Dioxolane, stabilized
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.