missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyluric Acid, TRC

Product Code. 30134123
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134123 1 g 1g
30005021 250 mg 250mg
30012477 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134123 Supplier Toronto Research Chemicals Supplier No. TRCD4797351G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 944-73-0
Molecular Formula C7H8N4O3
Molecular Weight (g/mol) 196.16
Synonym 7,9-Dihydro-1,3-dimethyl-1H-purine-2,6,8(3H)-trione, 1,3-Dimethyl Uric Acid;
IUPAC Name 1,3-dimethyl-7,9-dihydropurine-2,6,8-trione
SMILES CN1C(=O)N(C)C2=C(NC(=O)N2)C1=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H8N4O3/c1-10-4-3(8-6(13)9-4)5(12)11(2)7(10)14/h1-2H3,(H2,8,9,13)
Formula Weight 196.0596
Percent Purity >95%
Chemical Name or Material 1,3-Dimethyluric Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.