missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyluric acid, MedChemExpress

Product Code. 16435457
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16435457 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16435457 Supplier Medchem Express Supplier No. HYW014993
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 1,3-Dimethyluric acid is a product of theophylline metabolism in man. 1,3-Dimethyluric acid is one of the purine components in urinary calculi.

1,3-Dimethyluric acid is a product of theophylline metabolism in man. 1,3-Dimethyluric acid is one of the purine components in urinary calculi.

Specifications

Chemical Name or Material 1,3-Dimethyluric acid
CAS 944-73-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C7H8N4O3
Quantity 50 mg
Solubility Information DMSO : 5 mg/mL (25.49 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(N1C)N(C)C2=C(NC(N2)=O)C1=O
Molecular Weight (g/mol) 196.16
Formula Weight 196.16
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.