missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethylbarbituric acid, 98%

Product Code. 10277683 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10277683 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10277683 Supplier Thermo Scientific Acros Supplier No. 393285000
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 769-42-6 | C6H8N2O3 | 156.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • 1,3-Dimethylbarbituric acid, 98% 500g Plastic Bottle Cas No. 769-42-6
TRUSTED_SUSTAINABILITY

Specifications

CAS 769-42-6
Melting Point 121°C to 123°C (literature)
Boiling Point 228.1°C ± 23.0°C
Packaging Glass Bottle
Molecular Formula C6H8N2O3
Quantity 500 g
InChI Key VVSASNKOFCZVES-UHFFFAOYSA-N
SMILES CN1C(=O)CC(=O)N(C)C1=O
IUPAC Name 1,3-dimethyl-1,3-diazinane-2,4,6-trione
Molecular Weight (g/mol) 156.14
Percent Purity 98%
Physical Form Powder
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,3-Dimethylbarbituric acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.