missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethylbarbituric acid, 98%

Product Code. 10573734 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10573734 25 g 25g
10106213 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10573734 Supplier Thermo Scientific Acros Supplier No. 393280250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 769-42-6 | C6H8N2O3 | 156.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 769-42-6
Molecular Formula C6H8N2O3
Molecular Weight (g/mol) 156.14
InChI Key VVSASNKOFCZVES-UHFFFAOYSA-N
IUPAC Name 1,3-dimethyl-1,3-diazinane-2,4,6-trione
SMILES CN1C(=O)CC(=O)N(C)C1=O

Specifications

Melting Point 122.0°C to 125.0°C
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility aq. soln.: 60 g/L (20°C). Other solubilities: soluble in alcohols and acetone
Formula Weight 156.14
Chemical Name or Material 1,3-Dimethylbarbituric acid
compliance-icons compliance-icons
Product Identifier
  • 1,3-Dimethylbarbituric acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.