missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, 97%

Product Code. p-7027966
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446037 5 g 5g
11456037 25 g 25g
11466037 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11446037 Supplier Thermo Scientific Alfa Aesar Supplier No. B25057.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1892-57-5 | C8H17N3 | 155.245 g/mol

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide is used as a carboxyl activating agent and activate phosphate groups in phospho mono and di esters. It is used in peptide synthesis, 3'-amino-3'-deoxyadenosine-5'-di- and triphosphates and in the preparation of antibodies like immunoconjugates. It plays a vital role for immobilization of large biomolecules in association with N-hydroxysuccinimide. It is also used in the acylation of phosphoranes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide is used as a carboxyl activating agent and activate phosphate groups in phospho mono and di esters. It is used in peptide synthesis, 3′-amino-3′-deoxyadenosine-5′-di- and triphosphates and in the preparation of antibodies like immunoconjugates. It plays a vital role for immobilization of large biomolecules in association with N-hydroxysuccinimide. It is also used in the acylation of phosphoranes.

Solubility
Soluble in water.

Notes
Air sensitive. Incompatible with strong acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1892-57-5
Molecular Formula C8H17N3
Molecular Weight (g/mol) 155.245
MDL Number MFCD00044916
InChI Key LMDZBCPBFSXMTL-UHFFFAOYSA-N
Synonym 1-3-dimethylaminopropyl-3-ethylcarbodiimide, edac, 1-ethyl-3-3-dimethylaminopropyl carbodiimide, n1-ethylimino methylene-n3,n3-dimethylpropane-1,3-diamine, unii-rj5ozg6i4a, edci, 1,3-propanediamine, n'-ethylcarbonimidoyl-n,n-dimethyl, n-3-dimethylaminopropyl-n'-ethylcarbodiimide, rj5ozg6i4a, ethyldimethylaminopropyl carbodiimide
PubChem CID 15908
IUPAC Name 3-(ethyliminomethylideneamino)-N,N-dimethylpropan-1-amine
SMILES CCN=C=NCCCN(C)C

Specifications

Density 0.885
Boiling Point 66°C to 68°C (1 mmHg)
Flash Point >110°C (230°F)
Linear Formula CH3CH2N=C=N(CH2)3N(CH3)2
Refractive Index 1.461
Quantity 5 g
UN Number UN2735
Beilstein 507429
Sensitivity Air and moisture sensitive
Solubility Information Soluble in water.
Formula Weight 155.24
Percent Purity 97%
Chemical Name or Material 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide
Show More Show Less
compliance-icons
Product Identifier
  • 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.