missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-5-aminoadamantane hydrochloride, 95%

Product Code. 10183481 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10183481 1 g Glass bottle 1g
10193481 10 g Glass bottle 10g
11935201 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10183481 Supplier Thermo Scientific Acros Supplier No. 298080010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41100-52-1 | C12H22ClN | 215.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41100-52-1
Molecular Formula C12H22ClN
Molecular Weight (g/mol) 215.77
MDL Number MFCD00214336
InChI Key LDDHMLJTFXJGPI-UHFFFAOYNA-N
Synonym memantine hydrochloride, memantine hcl, 3,5-dimethyladamantan-1-amine hydrochloride, 3,5-dimethyl-1-adamantanamine hydrochloride, namenda, akatinol, axura, namenda xr, memantine.hcl
PubChem CID 181458
ChEBI CHEBI:64323
IUPAC Name 3,5-dimethyladamantan-1-amine;hydrochloride
SMILES [H+].[Cl-].CC12CC3CC(C)(C1)CC(N)(C3)C2

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Merck Index 15,5897
Solubility Information Solubility in water: soluble
Formula Weight 215.77
Percent Purity 99%
Physical Form Fine Powder
Chemical Name or Material 1, 3-Dimethyl-5-aminoadamantane hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.