Learn More
1,3-Dimethyl-5-aminoadamantane hydrochloride, 95%, Thermo Scientific Chemicals
£53.10 - £1023.66
Chemical Identifiers
CAS | 41100-52-1 |
---|---|
Molecular Formula | C12H22ClN |
Molecular Weight (g/mol) | 215.77 |
MDL Number | MFCD00214336 |
InChI Key | LDDHMLJTFXJGPI-UHFFFAOYNA-N |
Synonym | memantine hydrochloride, memantine hcl, 3,5-dimethyladamantan-1-amine hydrochloride, 3,5-dimethyl-1-adamantanamine hydrochloride, namenda, akatinol, axura, namenda xr, memantine.hcl |
PubChem CID | 181458 |
ChEBI | CHEBI:64323 |
IUPAC Name | 3,5-dimethyladamantan-1-amine;hydrochloride |
SMILES | [H+].[Cl-].CC12CC3CC(C)(C1)CC(N)(C3)C2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10183481
|
Thermo Scientific Acros
298080010 |
1 g | Glass bottle |
£53.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10193481
|
Thermo Scientific Acros
298080100 |
10 g | Glass bottle |
£300.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11935201
|
Thermo Scientific Acros
298080500 |
50 g | Glass Bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
41100-52-1 | |
215.77 | |
LDDHMLJTFXJGPI-UHFFFAOYNA-N | |
181458 | |
3,5-dimethyladamantan-1-amine;hydrochloride |
C12H22ClN | |
MFCD00214336 | |
memantine hydrochloride, memantine hcl, 3,5-dimethyladamantan-1-amine hydrochloride, 3,5-dimethyl-1-adamantanamine hydrochloride, namenda, akatinol, axura, namenda xr, memantine.hcl | |
CHEBI:64323 | |
[H+].[Cl-].CC12CC3CC(C)(C1)CC(N)(C3)C2 |
Specifications
41100-52-1 | |
Authentic | |
C12H22ClN | |
15,5897 | |
Solubility in water: soluble | |
[H+].[Cl-].CC12CC3CC(C)(C1)CC(N)(C3)C2 | |
215.77 | |
CHEBI:64323 | |
99% | |
1, 3-Dimethyl-5-aminoadamantane hydrochloride |
White | |
99% | |
MFCD00214336 | |
memantine hydrochloride, memantine hcl, 3,5-dimethyladamantan-1-amine hydrochloride, 3,5-dimethyl-1-adamantanamine hydrochloride, namenda, akatinol, axura, namenda xr, memantine.hcl | |
LDDHMLJTFXJGPI-UHFFFAOYNA-N | |
3,5-dimethyladamantan-1-amine;hydrochloride | |
181458 | |
215.77 | |
Fine Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protec
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 255-219-6
RUO – Research Use Only