missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-5-nitroadamantane, TRC

Product Code. 30370107
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30370107 100 mg -
30372937 1 g 1g
30382634 500 mg -
3 options
This item is not returnable. View return policy

Product Code. 30370107

Brand: Toronto Research Chemicals TRCD476665100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6588-68-7
Formula Weight 209.1416
InChI Formula InChI=1S/C12H19NO2/c1-10-3-9-4-11(2,6-10)8-12(5-9,7-10)13(14)15/h9H,3-8H2,1-2H3/t9-,10+,11-,12-
IUPAC Name (1S,3R)-1,3-dimethyl-5-nitroadamantane
Molecular Formula C12H19NO2
Molecular Weight (g/mol) 209.28
SMILES O=[N+]([O-])[C@@]1(C2)C[C@]3(C)C[C@H]2C[C@@](C1)(C)C3
Synonym 1,3-Dimethyl-5-nitroadamantane
Chemical Name or Material 1,3-Dimethyl-5-nitroadamantane

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.