missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone, 99+%, ACROS Organics™

Product Code. 15259636 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
AcroSeal™ Glass bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15259636 1 L AcroSeal™ Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15259636 Supplier Thermo Scientific Acros Supplier No. 433560010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Coupling reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 7226-23-5
Assay Percent Range 99+%
Packaging AcroSeal™ Glass bottle
Molecular Formula C6H12N2O
Quantity 1 L
Synonym 1,3-dimethyl-3,4,5,6-tetrahydro-2 1h-pyrimidinone, 1,3-dimethyltetrahydropyrimidin-2 1h-one, dmpu, dimethylpropyleneurea, 2 1h-pyrimidinone, tetrahydro-1,3-dimethyl, n,n'-dimethylpropyleneurea, n,n'-dimethyltrimethyleneurea, ccris 4322, n,n'-dimethylpropylene urea, tetrahydro-1,3-dimethyl-2 1h pyrimidine
InChI Key GUVUOGQBMYCBQP-UHFFFAOYSA-N
SMILES CN1CCCN(C1=O)C
IUPAC Name 1,3-dimethyl-1,3-diazinan-2-one
Molecular Weight (g/mol) 128.175
PubChem CID 81646
Formula Weight 128.17
Grade Extra Dry
Chemical Name or Material 1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,3-Dimethyl-3,4,5,6-tetrahydro-2(1H)-pyrimidinone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H361f-Suspected of damaging fertility.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.