missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dimethyl-1H-thieno[2,3-c]pyrazole-5-carboxylic acid, 95+%, Thermo Scientific™

Product Code. 10079141
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10079141 250 mg Amber Glass Bottle 250mg
10089141 1 g Amber Glass Bottle 1g
10507864 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10079141 Supplier Thermo Scientific Maybridge Supplier No. CC22901CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 25252-46-4
Molecular Formula C8H8N2O2S
Molecular Weight (g/mol) 196.224
InChI Key QRANSYHQSVJLHX-UHFFFAOYSA-N
Synonym 1,3-dimethyl-1h-thieno 2,3-c pyrazole-5-carboxylic acid, 1,3-dimethylthieno 2,3-c pyrazole-5-carboxylic acid, 1h-thieno 2,3-c pyrazole-5-carboxylic acid, 1,3-dimethyl, 1h-thieno 2,3-c pyrazole-5-carboxylicacid, 1,3-dimethyl, 4fzj, 1,3-dimethyl-1h-thieno 2,3-c pyrazole-5-carboxylicacid, unavailable-see asdi_catno 500021541, 5-carboxy-1,3-dimethyl-1h-thieno 2,3-c pyrazole, 1,3-dimethylthiopheno 3,2-d pyrazole-5-carboxylic acid, 1,3-dimethyl-1h-thieno 2,3-c-pyrazole-5-carboxylic acid
PubChem CID 2776541
IUPAC Name 1,3-dimethylthieno[2,3-c]pyrazole-5-carboxylic acid
SMILES CC1=NN(C2=C1C=C(S2)C(=O)O)C

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 196.23
Percent Purity 95%
Chemical Name or Material 1, 3-Dimethyl-1H-thieno[2, 3-c]pyrazole-5-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.