missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Specifications
Assay Percent Range | 97% |
Quantity | 10 g |
Formula Weight | 140.14 |
Percent Purity | 97% |
Chemical Name or Material | 1, 3-Dimethyl-1H-pyrazole-5-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'