missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diisopropenylbenzene, TRC

Product Code. 30007537
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007537 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30007537 Supplier Toronto Research Chemicals Supplier No. TRCD6791405ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3748-13-8
Molecular Formula C12H14
Quantity 5 mL
InChI Formula InChI=1S/C12H14/c1-9(2)11-6-5-7-12(8-11)10(3)4/h5-8H,1,3H2,2,4H3
SMILES C=C(C)C1=CC(=CC=C1)C(=C)C
IUPAC Name 1,3-diisopropenylbenzene
Molecular Weight (g/mol) 158.24
Formula Weight 158.1096
Chemical Name or Material 1,3-Diisopropenylbenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.