missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diiodopropane, TRC

Product Code. 30371419
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
30371419 10 mg 10mg
1 options
This item is not returnable. View return policy

Product Code. 30371419

Brand: Toronto Research Chemicals TRCD45571910MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Diiodopropane
CAS 627-31-6
Molecular Formula C3H6I2
Quantity 10 mg
Synonym NSC 35647, Trimethylene Diiodide, alpha,ω-Diiodopropane
InChI Formula InChI=1S/C3H6I2/c4-2-1-3-5/h1-3H2
SMILES ICCCI
IUPAC Name 1,3-diiodopropane
Molecular Weight (g/mol) 295.89
Formula Weight 295.8559
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.