missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diiodopropane, 99%, stabilized

Product Code. 10226990 Shop All Thermo Scientific Chemicals Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
missing translation for 'unitSize'
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10226990 25 mL Glass bottle 25mL
15740839 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 10226990 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 218600250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 627-31-6 | C3H6I2 | 295.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-31-6
Molecular Formula C3H6I2
Molecular Weight (g/mol) 295.88
MDL Number MFCD00001097
InChI Key AAAXMNYUNVCMCJ-UHFFFAOYSA-N
Synonym trimethylene diiodide, propane, 1,3-diiodo, trimethylene iodide, propane,3-diiodo, pubchem3858, 1,3-bis iodanyl propane, acmc-1av5p, 4-01-00-00228 beilstein handbook reference, 1,3-diiodopropane, contains copper as stabilizer
PubChem CID 12314
IUPAC Name 1,3-diiodopropane
SMILES C(CI)CI

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Pink to Yellow
Density 2.5760g/mL
Boiling Point 111.0°C to 113.0°C (31.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula I(CH2)3I
Refractive Index 1.6405 to 1.6425
Quantity 25 mL
Beilstein 01, 115
Specific Gravity 2.576
Solubility Information Solubility in water: insoluble in water.
Stabilizer copper
Formula Weight 295.88
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1,3-Diiodopropane, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1,3-Diiodopropane, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.