missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diiodopropane, 98+%, stab. with copper

Product Code. 11483314
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483314 50 g 50g
11493314 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483314 Supplier Thermo Scientific Alfa Aesar Supplier No. A10616.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 627-31-6 | C3H6I2 | 295.89 g/mol

1,3-Diiodopropane is used as a reagent for organic synthesis. 1, 3-Diiodopropane is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Diiodopropane is used as a reagent for organic synthesis. 1, 3-Diiodopropane is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Stable, but light sensitive - store in the dark. Incompatible with strong bases, strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-31-6
Molecular Formula C3H6I2
Molecular Weight (g/mol) 295.89
MDL Number MFCD00001097
InChI Key AAAXMNYUNVCMCJ-UHFFFAOYSA-N
Synonym trimethylene diiodide, propane, 1,3-diiodo, trimethylene iodide, propane,3-diiodo, pubchem3858, 1,3-bis iodanyl propane, acmc-1av5p, 4-01-00-00228 beilstein handbook reference, 1,3-diiodopropane, contains copper as stabilizer
PubChem CID 12314
IUPAC Name 1,3-diiodopropane
SMILES C(CI)CI

Specifications

Melting Point -20°C
Density 2.565
Boiling Point 224°C
Flash Point >110°C (230°F)
Refractive Index 1.6415
Quantity 50 g
Beilstein 1731117
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 295.89
Percent Purity ≥98%
Chemical Name or Material 1,3-Diiodopropane, Stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.