missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diiodobenzene, 98+%

Product Code. 11398406
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398406 5 g 5g
11308416 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398406 Supplier Thermo Scientific Alfa Aesar Supplier No. L17445.06
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 626-00-6 | C6H4I2 | 329.907 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Insoluble in water. Soluble in hot methanol.

Notes
Light sensitive. Store in cool dry conditions. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-00-6
Molecular Formula C6H4I2
Molecular Weight (g/mol) 329.907
MDL Number MFCD00041731
InChI Key SFPQFQUXAJOWNF-UHFFFAOYSA-N
Synonym m-diiodobenzene, benzene, 1,3-diiodo, benzene, m-diiodo, pubchem3833, 1,3-di iodo benzene, 1,3-diiodobenzene, acmc-1b21u, ksc358c5j, 1,3-diiodo-benzen;benzene, 1,3-diiodo-;benzene, m-diiodo-;benzene,1,3-diiodo-;benzene,m-diiodo
PubChem CID 12270
IUPAC Name 1,3-diiodobenzene
SMILES C1=CC(=CC(=C1)I)I

Specifications

Melting Point 35°C to 37°C
Density 2.47
Boiling Point 285°C
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1904540
Sensitivity Light sensitive
Solubility Information Insoluble in water. Soluble in hot methanol.
Formula Weight 329.91
Percent Purity ≥98%
Chemical Name or Material 1,3-Diiodobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.