missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dihydroxynaphthalene, 98%

Product Code. 11443338
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443338 25 g 25g
11423338 1 g 1g
11433338 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11443338 Supplier Thermo Scientific Alfa Aesar Supplier No. A17739.14
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 132-86-5 | C10H8O2 | 160.172 g/mol

It is used in the naphthoresorcinol test for uronic acids of Tollens and Rorive, a blue color can be extracted with ethanol after heating of uronic acids with naphthoresorcinol in hydrochloric acid. It is also employed as a reagent for various medical tests. It can also be used as a spot test reagent for glycerate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the naphthoresorcinol test for uronic acids of Tollens and Rorive, a blue color can be extracted with ethanol after heating of uronic acids with naphthoresorcinol in hydrochloric acid. It is also employed as a reagent for various medical tests. It can also be used as a spot test reagent for glycerate.

Solubility
Soluble in ethanol (50 mg/ml), and water.

Notes
Light Sensitive. Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry and well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132-86-5
Molecular Formula C10H8O2
MDL Number MFCD00003965
InChI Key XOOMNEFVDUTJPP-UHFFFAOYSA-N
Synonym 1,3-naphthalenediol, 1,3-dihydroxynaphthalene, naphthoresorcinol, naphthoresoucinol, 3-hydroxybenzocyclohexadien-1-one, naphthoresorcin, ccris 7896, unii-5x457yew8y, acmc-1bvwb, naphthalenediol-1,3
PubChem CID 8601
IUPAC Name naphthalene-1,3-diol

Specifications

Melting Point 123°C to 126°C
Quantity 25 g
Beilstein 2044002
Sensitivity Light sensitive
Merck Index 14,6396
Solubility Information Soluble in ethanol (50 mg/ml),and water.
SMILES OC1=CC(O)=C2C=CC=CC2=C1
Molecular Weight (g/mol) 160.17
Formula Weight 160.17
Percent Purity 98%
Chemical Name or Material 1,3-Dihydroxynaphthalene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.