Learn More
1,3-Dihydroxyacetone dimer, 97%, Thermo Scientific Chemicals
£20.60 - £235.00
Chemical Identifiers
CAS | 62147-49-3 |
---|---|
Molecular Formula | C3H6O3 |
Molecular Weight (g/mol) | 90.08 |
MDL Number | MFCD00051019 |
InChI Key | RXKJFZQQPQGTFL-UHFFFAOYSA-N |
Synonym | 1,3-dihydroxyacetone dimer, 1,3-dihydroxypropan-2-one dimer, 2,5-bis hydroxymethyl-1,4-dioxane-2,5-diol, 2,5-dihydroxy-1,4-dioxane-2,5-dimethanol, dihydroxyacetone dimer, 2,5-dihydroxydioxane-2,5-dimethanol, 1,4-dioxane-2,5-dimethanol, 2,5-dihydroxy, acmc-20aoav, pubchem4073 |
PubChem CID | 4180364 |
SMILES | OCC(=O)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11462257
|
Thermo Scientific Alfa Aesar
A14189.18 |
50 g |
£20.60
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11472257
|
Thermo Scientific Alfa Aesar
A14189.30 |
250 g |
£74.90
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11452257
|
Thermo Scientific Alfa Aesar
A14189.0B |
1000 g |
£235.00
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1,3-Dihydroxyacetone dimer is used as a catalytic agent and petrochemical additive. It is also used as a building block in organic synthesis. Further, it serves as a reagent in three-carbon nucleophilic or electrophilic component in various reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1,3-Dihydroxyacetone dimer is used as a catalytic agent and petrochemical additive. It is also used as a building block in organic synthesis. Further, it serves as a reagent in three-carbon nucleophilic or electrophilic component in various reactions.
Solubility
Soluble in ethanol, ethyl ether, acetone. Slightly soluble in water. Insoluble in ligroin.
Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
Chemical Identifiers
62147-49-3 | |
90.08 | |
RXKJFZQQPQGTFL-UHFFFAOYSA-N | |
4180364 |
C3H6O3 | |
MFCD00051019 | |
1,3-dihydroxyacetone dimer, 1,3-dihydroxypropan-2-one dimer, 2,5-bis hydroxymethyl-1,4-dioxane-2,5-diol, 2,5-dihydroxy-1,4-dioxane-2,5-dimethanol, dihydroxyacetone dimer, 2,5-dihydroxydioxane-2,5-dimethanol, 1,4-dioxane-2,5-dimethanol, 2,5-dihydroxy, acmc-20aoav, pubchem4073 | |
OCC(=O)CO |
Specifications
∼75°C to 82°C | |
C3H6O3 | |
112910 | |
1,3-dihydroxyacetone dimer, 1,3-dihydroxypropan-2-one dimer, 2,5-bis hydroxymethyl-1,4-dioxane-2,5-diol, 2,5-dihydroxy-1,4-dioxane-2,5-dimethanol, dihydroxyacetone dimer, 2,5-dihydroxydioxane-2,5-dimethanol, 1,4-dioxane-2,5-dimethanol, 2,5-dihydroxy, acmc-20aoav, pubchem4073 | |
RXKJFZQQPQGTFL-UHFFFAOYSA-N | |
2,5-bis(hydroxymethyl)-1,4-dioxane-2,5-diol | |
4180364 | |
97% |
62147-49-3 | |
MFCD00051019 | |
14,3182 | |
Soluble in ethanol,ethyl ether,acetone. Slightly soluble in water. Insoluble in ligroin. | |
OCC(=O)CO | |
90.08 | |
180.16 | |
1,3-Dihydroxyacetone dimer |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 612-947-2
missing translation for 'tsca' : No
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only