missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dihydro-1,3-dioxo-5-isobenzofurancarboxylic Acid Phenylmethyl Ester, TRC

Product Code. 30386287
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386287 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386287 Supplier Toronto Research Chemicals Supplier No. TRCD450185100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Dihydro-1,3-dioxo-5-isobenzofurancarboxylic Acid Phenylmethyl Ester
CAS 29638-13-9
Molecular Formula C16H10O5
Quantity 100 mg
Synonym 1,2,4-Benzenetricarboxylic Acid, Cyclic 1,2-Anhydride, Benzyl Ester
InChI Formula InChI=1S/C16H10O5/c17-14(20-9-10-4-2-1-3-5-10)11-6-7-12-13(8-11)16(19)21-15(12)18/h1-8H,9H2
SMILES O=C(OCc1ccccc1)c2ccc3C(=O)OC(=O)c3c2
IUPAC Name benzyl 1,3-dioxo-2-benzofuran-5-carboxylate
Molecular Weight (g/mol) 282.25
Formula Weight 282.0528
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.