missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Dihydro-1-(1,2,3,6-tetrahydro-4-pyridinyl)-2H-benzimidazol-2-one, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | 1-(1,2,3,6-Tetrahydropyridin-4-yl)-1,3-dihydro-2H-benzimidazol-2-one |
| CAS | 2147-83-3 |
| Molecular Formula | C12 H13 N3 O |
| Quantity | 250 mg |
| Synonym | 1-(1,2,3,6-Tetrahydropyridin-4-yl)-1,3-dihydro-2H-benzimidazol-2-one, Droperidol Imp. A (EP) |
| InChI Formula | InChI=1S/C12H13N3O/c16-12-14-10-3-1-2-4-11(10)15(12)9-5-7-13-8-6-9/h1-5,13H,6-8H2,(H,14,16) |
| SMILES | O=C1Nc2ccccc2N1C3=CCNCC3 |
| IUPAC Name | 3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-benzimidazol-2-one |
| Molecular Weight (g/mol) | 215.25 |
| Formula Weight | 215.1059 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction