missing translation for 'onlineSavingsMsg'
Learn More

1,3-Difluorobenzene, 99%

Product Code. 11446987
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446987 10 g 10g
11456987 50 g 50g
11466987 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11446987 Supplier Thermo Scientific Alfa Aesar Supplier No. A14387.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 372-18-9 | C6H4F2 | 114.10 g/mol

It is used in the laser induced fluorencence matrix study of 1,3-difluorobenzene radical cation. It is also used synthesis of 1,2,3-triazole containing fluconazole analogues.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the laser induced fluorencence matrix study of 1,3-difluorobenzene radical cation. It is also used synthesis of 1,2,3-triazole containing fluconazole analogues.

Solubility
Soluble in water at 20°C 1.1g/L.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-18-9
Molecular Formula C6H4F2
Molecular Weight (g/mol) 114.10
MDL Number MFCD00000327
InChI Key UEMGWPRHOOEKTA-UHFFFAOYSA-N
Synonym m-difluorobenzene, benzene, 1,3-difluoro, benzene, m-difluoro, 2,4-difluorobenzene, meta-difluorobenzene, 1,3-difluoro benzene, unii-vo3c26woc2, 1,3-difluorbenzol, vo3c26woc2, 1,3difluorobenzene
PubChem CID 9741
ChEBI CHEBI:38584
IUPAC Name 1,3-difluorobenzene
SMILES FC1=CC(F)=CC=C1

Specifications

Melting Point -59°C
Density 1.16
Boiling Point 82°C to 83°C
Flash Point 2°C (36°F)
Odor Characteristic
Refractive Index 1.438
Quantity 10 g
UN Number UN1993
Beilstein 1904537
Solubility Information Soluble in water at 20°C 1.1g/L.
Formula Weight 114.09
Percent Purity 99%
Chemical Name or Material 1,3-Difluorobenzene
Show More Show Less
compliance-icons
Product Identifier
  • 1,3-Difluorobenzene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Supplemental information
  • MIXTURE LIST-Contain: 1,3-Difluorobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.