missing translation for 'onlineSavingsMsg'
Learn More

1,3-Difluoroacetone, 98%

Product Code. 15418027
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418027 1 g 1g
15428027 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418027 Supplier Thermo Scientific Alfa Aesar Supplier No. H51145.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 453-14-5 | C3H4F2O | 94.061 g/mol

1,3-Difluoroacetone is used as pharmaceutical intermediate and for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Difluoroacetone is used as pharmaceutical intermediate and for chemical research.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 453-14-5
Molecular Formula C3H4F2O
Molecular Weight (g/mol) 94.061
MDL Number MFCD07368665
InChI Key HKIPCXRNASWFRU-UHFFFAOYSA-N
Synonym 1,3-difluoroacetone, 2-propanone, 1,3-difluoro, 1,3-difluoro-2-propanone, ch2f 2co, 1,3-bis fluoranyl propan-2-one, fluoromethyl ketone, sym-difluoroacetone, sgqdlibrbrwsp@, 1, 3-difluoroacetone, 1,3-difluoro acetone
PubChem CID 228397
IUPAC Name 1,3-difluoropropan-2-one
SMILES C(C(=O)CF)F

Specifications

Boiling Point 101°C to 102°C
Quantity 1 g
UN Number UN2929
Formula Weight 94.06
Percent Purity 98%
Chemical Name or Material 1,3-Difluoroacetone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.