missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Diethylbenzene, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
2 mL
10 mL
Description
1,3-Diethylbenzene is used as an intermediate and solvent. It is also used in the preparation of divinyl benzene. Further, it is used in powderless etching.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 0.86 |
Boiling Point | 182°C |
Flash Point | 55°C (131°F) |
Assay Percent Range | >97% |
Refractive Index | 1.496 |
Quantity | 10 mL |
UN Number | UN2049 |
Solubility Information | Miscible with ethanol,benzene,carbon tetrachloride,ethyl ether and acetone. Immiscible with water and diethylbenzene. |
Formula Weight | 134.22 |
Percent Purity | ≥97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'