missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diethyl ester 2-[3-[(3aS,4S,6aR)-hexahydro-2-oxo-1,3-bis(phenylmethyl)-1H-thieno[3,4-d]imidazol-4-yl]propyl] Propanedioic acid, TRC

Product Code. 30371103
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30371103 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30371103 Supplier Toronto Research Chemicals Supplier No. TRCD4561002.5MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Diethyl 4-[(3aS,4S,6aR)-1,3-Dibenzyl-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl]butane-1,1-dicarboxylate
CAS 101469-35-6
Molecular Formula C29 H36 N2 O5 S
Quantity 2.5 mg
Synonym Propanedioic acid, 2-[3-[(3aS,4S,6aR)-hexahydro-2-oxo-1,3-bis(phenylmethyl)-1H-thieno[3,4-d]imidazol-4-yl]propyl]-, 1,3-diethyl ester, Propanedioic acid, [3-[(3aS,4S,6aR)-hexahydro-2-oxo-1,3-bis(phenylmethyl)-1H-thieno[3,4-d]imidazol-4-yl]propyl]-, diethyl ester (9CI), Propanedioic acid, [3-[hexahydro-2-oxo-1,3-bis(phenylmethyl)-1H-thieno[3,4-d]imidazol-4-yl]propyl]-, diethyl ester, [3aS-(3aα,4β,6aα)]-, 1H-Thieno[3,4-d]imidazole, propanedioic acid deriv., Diethyl 4-[(3aS,4S,6aR)-1,3-Dibenzyl-2-oxohexahydro-1H-thieno[3,4-d]imidazol-4-yl]butane-1,1-dicarboxylate
InChI Formula InChI=1S/C29H36N2O5S/c1-3-35-27(32)23(28(33)36-4-2)16-11-17-25-26-24(20-37-25)30(18-21-12-7-5-8-13-21)29(34)31(26)19-22-14-9-6-10-15-22/h5-10,12-15,23-26H,3-4,11,16-20H2,1-2H3/t24-,25-,26-/m0/s1
SMILES CCOC(=O)C(CCC[C@@H]1SC[C@H]2[C@@H]1N(Cc3ccccc3)C(=O)N2Cc4ccccc4)C(=O)OCC
IUPAC Name diethyl 2-[3-[(3aS,4S,6aR)-1,3-dibenzyl-2-oxo-3a,4,6,6a-tetrahydrothieno[3,4-d]imidazol-4-yl]propyl]propanedioate
Molecular Weight (g/mol) 524.672
Formula Weight 524.234
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.