missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diethyl-2-thiourea, TRC

Product Code. 30380801
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30380801 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380801 Supplier Toronto Research Chemicals Supplier No. TRCD4452935G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N'-Diethylthiourea
CAS 105-55-5
Molecular Formula C5 H12 N2 S
Quantity 5 g
Synonym Urea, 1,3-diethyl-2-thio- (6CI,8CI), N,N'-Diethylthiourea, 1,3-Diethyl-2-thiourea, 1,3-Diethylthiourea, Accel EUR, Accelerator DETU, Accelerator EUR, Diethylthiourea, N,N'-Diethylthiocarbamide, NSC 3507, Nocceler EUR, Pennzone E, Rhenogran DETU, Robac DETU, Sanceler EUR, Thiate H, U 15030, N,N'-Diethylthiourea
InChI Formula InChI=1S/C5H12N2S/c1-3-6-5(8)7-4-2/h3-4H2,1-2H3,(H2,6,7,8)
SMILES CCNC(=S)NCC
IUPAC Name 1,3-diethylthiourea
Molecular Weight (g/mol) 132.23
Formula Weight 132.0721
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.