missing translation for 'onlineSavingsMsg'
Learn More

1,3-Diethoxybenzene, 95%

Product Code. 11493744
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493744 50 g 50g
11483744 10 g 10g
11403754 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493744 Supplier Thermo Scientific Alfa Aesar Supplier No. A11338.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2049-73-2 | C10H14O2 | 166.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2049-73-2
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00015145
InChI Key MKGFYMKFBCWNCP-UHFFFAOYSA-N
Synonym m-diethoxybenzene, benzene, 1,3-diethoxy, resorcinol diethyl ether, resorcindiathylather, benzene,3-diethoxy, acmc-1cdp4, benzene, 1,3-diethoxy-9ci
PubChem CID 74899
IUPAC Name 1,3-diethoxybenzene
SMILES CCOC1=CC(=CC=C1)OCC

Specifications

Melting Point 11°C to 12°C
Density 1.01
Boiling Point 234°C to 236°C
Refractive Index 1.5095
Quantity 50 g
Beilstein 2517730
Formula Weight 166.22
Percent Purity 95%
Chemical Name or Material 1,3-Diethoxybenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.