missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dicyclohexylimidazolium tetrafluoroborate, 97%

Product Code. 15457677
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15457677 5 g 5g
15447677 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15457677 Supplier Thermo Scientific Alfa Aesar Supplier No. H36633.06
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 286014-38-8 | C15H25BF4N2 | 320.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 286014-38-8
Molecular Formula C15H25BF4N2
Molecular Weight (g/mol) 320.18
MDL Number MFCD04973310
InChI Key CQHXJIHJFMBBQA-UHFFFAOYSA-N
Synonym 1,3-dicyclohexyl-1h-imidazol-3-ium tetrafluoroborate, 1,3-dicyclohexylimidazolium tetrafluoroborate, 1,3-dicyclohexyl-imidazolium tetrafluoroborate, 1,3-bis cyclohexyl imidazolium tetrafluoroborate, 1,3-dicyclohexyl-1??-imidazol-1-ylium tetrafluoroborate, acmc-1cq4s, 1,3-dicyclohexylimidazoliumtetrafluoroboratesalt, 1,3-dicyclohexylimidazolium tetrafluoroborate salt
PubChem CID 16218169
IUPAC Name 1,3-dicyclohexyl-3H-1λ⁵-imidazol-1-ylium; tetrafluoroboranuide
SMILES F[B-](F)(F)F.C1CCC(CC1)N1C=C[N+](=C1)C1CCCCC1

Specifications

Melting Point 173°C to 177°C
Color Brown to White
Quantity 5 g
Formula Weight 320.18
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 1,3-Dicyclohexylimidazolium tetrafluoroborate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.