missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dicyclohexylbenzimidazolium chloride, 95%

Product Code. 11916661 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11916661 500 mg 500mg
11906661 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11916661 Supplier Thermo Scientific Acros Supplier No. 435095000
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1034449-15-4 | C19H27ClN2 | 318.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 1034449-15-4
Molecular Formula C19H27ClN2
Molecular Weight (g/mol) 318.89
MDL Number MFCD08705248
InChI Key JACUDTKAZHCIPU-UHFFFAOYSA-M
Synonym 1,3-dicyclohexylbenzimidazolium chloride, 1,3-dicyclohexyl-1??,3-benzodiazol-1-ylium chloride, 1,3-dicyclohexyl-1h-benzimidazol-3-ium chloride, 1,3-dicyclohexyl-1h-benzo d imidazol-3-ium chloride, 1,3-dicyclohexyl-2h-1,3-benzodiazol-2-ylium chloride
PubChem CID 16218267
SMILES [Cl-].C1CCC(CC1)N1C=[N+](C2CCCCC2)C2=CC=CC=C12

Especificaciones

CAS Min % 94.0
CAS Max % 100.0
Melting Point 209.9°C to 211.1°C
Color Tan to White
Elemental Analysis 67.30 to 75.90% C, 8.25 to 9.31% N
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass Bottle
Quantity 500 mg
IUPAC Name 1,3-dicyclohexylbenzimidazol-3-ium;chloride
Formula Weight 318.89
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 1, 3-Dicyclohexylbenzimidazolium chloride
Mostrar más Mostrar menos
compliance-icons
Nombre del producto
  • 1,3-Dicyclohexylbenzimidazolium chloride
Palabra de advertencia
  • Warning
Clasificación
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Consejos de prudencia
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.