missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dicyanobenzene, 98%

Product Code. 10134370 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1kg
250g
5g
This item is not returnable. View return policy

Product Code. 10134370

Brand: Thermo Scientific Acros 163660050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 626-17-5 | C8H4N2 | 128.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-17-5
Molecular Formula C8H4N2
Molecular Weight (g/mol) 128.13
MDL Number MFCD00001795
InChI Key LAQPNDIUHRHNCV-UHFFFAOYSA-N
Synonym isophthalonitrile, 1,3-dicyanobenzene, isophthalodinitrile, 1,3-benzenedicarbonitrile, m-dicyanobenzene, 3-cyanobenzonitrile, m-benzenedinitrile, isoftalodinitril, 1,3-benzodinitrile, m-phthalodinitrile
PubChem CID 12276
ChEBI CHEBI:38218
IUPAC Name benzene-1,3-dicarbonitrile
SMILES N#CC1=CC(=CC=C1)C#N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 161°C to 165°C
Color Tan to White
Boiling Point 288°C
Flash Point >150°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(CN)2
Quantity 5 g
Beilstein 09, 836
Solubility Information Solubility in water: 0.7g/l (20°C). Other solubilities: 100g/l in acetone (20°C),insoluble in ether,acetone,hot ligroin,very soluble in alcohol,benzene and chloroform
Formula Weight 128.13
Percent Purity 98%
Physical Form Flakes
Chemical Name or Material 1, 3-Dicyanobenzene, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1,3-Dicyanobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.