missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloropropane, 99%, Thermo Scientific Chemicals

Catalog Number 10315940
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
142-28-9
C3H6Cl2
112.99
MFCD00000999
YHRUOJUYPBUZOS-UHFFFAOYSA-N
trimethylene dichloride, propane, 1,3-dichloro, trimethylene chloride, 1,3-dichloro-propane, unii-aj1hq2gucp, ccris 9220, ch2clch2ch2cl, aj1hq2gucp, cl ch2 3cl, 1,3-dichlorpropan
8881
1,3-dichloropropane
C(CCl)CCl
This item is not returnable. View return policy
142-28-9
C3H6Cl2
112.99
MFCD00000999
YHRUOJUYPBUZOS-UHFFFAOYSA-N
trimethylene dichloride, propane, 1,3-dichloro, trimethylene chloride, 1,3-dichloro-propane, unii-aj1hq2gucp, ccris 9220, ch2clch2ch2cl, aj1hq2gucp, cl ch2 3cl, 1,3-dichlorpropan
8881
1,3-dichloropropane
C(CCl)CCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -99.0°C
Color Colorless
Density 1.1900g/mL
Boiling Point 120.0°C to 122.0°C
Flash Point 20°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula Cl(CH2)3Cl
Refractive Index 1.4471 to 1.4491
Quantity 50 mL
Beilstein 01, 105
Specific Gravity 1.19
Solubility Information Solubility in water: 0.8g/L (20°C). Other solubilities: freely soluble in ethanol and ether
Viscosity 1.1 mPa.s (20°C)
Formula Weight 112.99
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material 1, 3-Dichloropropane, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,3-Dichloropropane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'