missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloroisoquinoline, 97%

Product Code. 10366874 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10366874 1 g 1g
10575714 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10366874 Supplier Thermo Scientific Acros Supplier No. 382410010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7742-73-6 | C9H5Cl2N | 198.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7742-73-6
Molecular Formula C9H5Cl2N
Molecular Weight (g/mol) 198.05
InChI Key BRGZEQXWZWBPJH-UHFFFAOYSA-N
PubChem CID 298625
IUPAC Name 1,3-dichloroisoquinoline
SMILES C1=CC=C2C(=C1)C=C(N=C2Cl)Cl

Specifications

Melting Point 120.0°C to 122.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Synonym isoquinoline, 1,3-dichloro, 1,3-dichloro-isoquinoline, pubchem7254, 1, 3-dichloroisoquinoline, acmc-209p9x, ksc493q3b, 1,3-dichloroisoquinoline
Formula Weight 198.05
Percent Purity 97%
Chemical Name or Material 1, 3-Dichloroisoquinoline
compliance-icons
Product Identifier
  • 1,3-Dichloroisoquinoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.