missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloroisopropyl-2,3-dichloropropyl Ether (>85%), TRC

Product Code. 30380043
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380043 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30380043 Supplier Toronto Research Chemicals Supplier No. TRCD4348551G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Dichloroisopropyl-2,3-dichloropropyl Ether
CAS 59440-90-3
Molecular Formula C6 H10 Cl4 O
Quantity 1 g
Synonym 1,3-Dichloroisopropyl-2,3-dichloropropyl Ether, Propane, 1,2-dichloro-3-[2-chloro-1-(chloromethyl)ethoxy]-, 1,2-Dichloro-3-[2-chloro-1-(chloromethyl)ethoxy]propane, 1,2-Dichloro-3-(1,3-dichloropropan-2-yloxy)propane, 1,3-Dichloroisopropyl-2,3-dichloropropyl ether, 1,3-Dichloroisopropyl-2,3-dichloropropyl Ether, Propane, 1,2-dichloro-3-[2-chloro-1-(chloromethyl)ethoxy]-, 1,2-Dichloro-3-[2-chloro-1-(chloromethyl)ethoxy]propane, 1,2-Dichloro-3-(1,3-dichloropropan-2-yloxy)propane, 1,3-Dichloroisopropyl-2,3-dichloropropyl ether
InChI Formula InChI=1S/C6H10Cl4O/c7-1-5(10)4-11-6(2-8)3-9/h5-6H,1-4H2
SMILES ClCC(Cl)COC(CCl)CCl
IUPAC Name 1,2-dichloro-3-(1,3-dichloropropan-2-yloxy)propane
Molecular Weight (g/mol) 239.96
Formula Weight 237.9486
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.