missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichlorobenzene, TRC

Product Code. 30378813
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
Unit Size:
500g
50g
Product Code. Quantity unitSize
30378813 500 g 500g
30372947 50 g 50g
30385255 250 g -
3 options
This item is not returnable. View return policy

Product Code. 30378813

Brand: Toronto Research Chemicals TRCD431930500G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-73-1
Formula Weight 145.969
InChI Formula InChI=1S/C6H4Cl2/c7-5-2-1-3-6(8)4-5/h1-4H
IUPAC Name 1,3-dichlorobenzene
Molecular Formula C6 H4 Cl2
Molecular Weight (g/mol) 147
SMILES Clc1cccc(Cl)c1
Synonym Benzene, m-dichloro- (8CI), 1,3-Dichlorobenzene, 2,4-Dichlorobenzene, 2,6-Dichlorobenzene, NSC 8754, m-Dichlorobenzene, m-Dichlorobenzol, m-Phenylene dichloride
Chemical Name or Material 1,3-Dichlorobenzene

Specifications

Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.