missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichlorobenzene, 98%

Product Code. 11434664
Change view
Click to view available options
Quantity:
250 g
500 g
1000 g
Unit Size:
1000g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434664 250 g 250g
11454230 500 g 500g
11424664 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434664 Supplier Thermo Scientific Alfa Aesar Supplier No. A13688.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 541-73-1 | C6H4Cl2 | 146.998 g/mol

1,3-Dichlorobenzene is used in medicine, and dyes. And it is also used to study the effect of solvent vapor pressure on the vertical nanowire of C60 molecules1. 1,3-Dichlorobenzene, a chlorinated aromatic hydrocarbon, can be detected in environmental samples using an advanced Fourier transform infrared spectroscopy-attenuated total reflectance (FTIR-ATR) sensor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dichlorobenzene is used in medicine, and dyes. And it is also used to study the effect of solvent vapor pressure on the vertical nanowire of C60 molecules1. 1,3-Dichlorobenzene, a chlorinated aromatic hydrocarbon, can be detected in environmental samples using an advanced Fourier transform infrared spectroscopy-attenuated total reflectance (FTIR-ATR) sensor.

Solubility
Difficult to mix.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-73-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 146.998
MDL Number MFCD00000573
InChI Key ZPQOPVIELGIULI-UHFFFAOYSA-N
Synonym m-dichlorobenzene, m-phenylene dichloride, m-dichlorobenzol, meta-dichlorobenzene, benzene, 1,3-dichloro, metadichlorobenzene, benzene, m-dichloro, m-phenylenedichloride, m-dcb, 3-dichlorobenzene
PubChem CID 10943
ChEBI CHEBI:36693
IUPAC Name 1,3-dichlorobenzene
SMILES C1=CC(=CC(=C1)Cl)Cl

Specifications

Melting Point -25°C
Density 1.288
Boiling Point 173°C
Flash Point 63°C (145°F)
Refractive Index 1.546
Quantity 250 g
UN Number UN3082
Beilstein 956618
Merck Index 14,3055
Solubility Information Difficult to mix.
Formula Weight 147
Percent Purity 98%
Chemical Name or Material 1,3-Dichlorobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.