missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichlorobenzene, 98%

Product Code. 10011810 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
250 mL
500 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
25mL
500mL
5mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10011810 1 L Glass bottle 1 L
10426990 5 mL Glass bottle 5mL
10635972 25 mL Glass bottle 25mL
10215530 250 mL Glass bottle 250mL
10377800 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10011810 Supplier Thermo Scientific Acros Supplier No. 151180010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 541-73-1 | C6H4Cl2 | 147 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-73-1
Molecular Formula C6H4Cl2
Molecular Weight (g/mol) 147
MDL Number MFCD00000573
InChI Key ZPQOPVIELGIULI-UHFFFAOYSA-N
Synonym m-dichlorobenzene, m-phenylene dichloride, m-dichlorobenzol, meta-dichlorobenzene, benzene, 1,3-dichloro, metadichlorobenzene, benzene, m-dichloro, m-phenylenedichloride, m-dcb, 3-dichlorobenzene
PubChem CID 10943
ChEBI CHEBI:36693
IUPAC Name 1,3-dichlorobenzene
SMILES C1=CC(=CC(=C1)Cl)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -24°C
Color Colorless to Yellow
Density 1.2800g/mL
Boiling Point 172°C to 173°C
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H4Cl2
Refractive Index 1.5450 to 1.547
Quantity 1 L
Beilstein 05, 202
Merck Index 15, 3064
Specific Gravity 1.28
Solubility Information Solubility in water: 0.0123g/100mL water (25°C). Other solubilities: soluble in organic solvents such as alcohol,,benzene,ether
Viscosity 1.045 mPa.s (23°C)
Formula Weight 147
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 3-Dichlorobenzene, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.