missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloroacetone, 96%

Product Code. 10694852 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
250g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10694852 50 g Glass bottle 50g
10624672 25 g Glass bottle 25g
10184330 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10694852 Supplier Thermo Scientific Acros Supplier No. 173630500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 534-07-6 | C3H4Cl2O | 126.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-07-6
Molecular Formula C3H4Cl2O
Molecular Weight (g/mol) 126.96
MDL Number MFCD00000937
InChI Key SUNMBRGCANLOEG-UHFFFAOYSA-N
Synonym 1,3-dichloroacetone, 1,3-dichloro-2-propanone, 1,3-dichloropropanone, s-dichloroacetone, sym-dichloroacetone, bis chloromethyl ketone, 2-propanone, 1,3-dichloro, acetone, 1,3-dichloro, chloromethyl ketone
PubChem CID 10793
IUPAC Name 1,3-dichloropropan-2-one
SMILES ClCC(=O)CCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 38°C to 42°C
Color White or Colorless
Density 1.3830g/mL
Boiling Point 172°C to 173°C
Flash Point 89°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2COCH2Cl
Quantity 50 g
Beilstein 01, 655
Fieser 02,109
Merck Index 15, 3061
Specific Gravity 1.383
Solubility Information Solubility in water: 27.9g/L water (20° c). Other solubilities: 464g/L toluene (20°C),soluble in alcohol,ether and hexane,slightly soluble in acetone,benzene,chloroform,and methanol
Formula Weight 126.97
Percent Purity 96%
Physical Form Crystals
Chemical Name or Material 1, 3-Dichloroacetone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1,3-Dichloroacetone
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
  • H311-Toxic in contact with skin.
  • H335-May cause respiratory irritation.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.