missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-5-iodobenzene, 99%

Product Code. 11434604
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11434604

Brand: Thermo Scientific Alfa Aesar A13192.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3032-81-3 | C6H3Cl2I | 272.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3032-81-3
Molecular Formula C6H3Cl2I
Molecular Weight (g/mol) 272.89
MDL Number MFCD00001047
InChI Key AATPRMRVLQZEHB-UHFFFAOYSA-N
Synonym 3,5-dichloroiodobenzene, 3,5-dichlorophenyl iodide, benzene, 1,3-dichloro-5-iodo, 1,3-dichloro-5-iodo-benzene, 3,5-dichloro-1-iodobenzene, pubchem3693, acmc-209hev, 3,5-dichloro iodobenzene, 3,5-dichloro-iodobenzene, 3,5-dichloro-iodo-benzene
PubChem CID 76424
IUPAC Name 1,3-dichloro-5-iodobenzene
SMILES ClC1=CC(I)=CC(Cl)=C1

Specifications

Melting Point 55°C to 59°C
Boiling Point 127°C (20 mmHg)
Quantity 5 g
Beilstein 2242455
Sensitivity Light sensitive
Formula Weight 272.9
Percent Purity 99%
Chemical Name or Material 1,3-Dichloro-5-iodobenzene
compliance-icons
Product Identifier
  • 3,5-Dichloroiodobenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 3,5-Dichloroiodobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.