missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-2-propanol, TRC

Product Code. 30138382
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
Unit Size:
1g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138382 50 g 50g
30132846 5 g 5g
30009021 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138382 Supplier Toronto Research Chemicals Supplier No. TRCD43580050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-23-1
Molecular Formula C3 H6 Cl2 O
Molecular Weight (g/mol) 128.99
Synonym 1,3-Dichloro-2-propanol, 1,3-Dichloro-2-hydroxypropane, 1,3-Dichlorohydrin, 1,3-Dichloroisopropanol, 1,3-Dichloroisopropyl alcohol, 2-Chloro-1-(chloromethyl)ethanol, Bis(chloromethyl)methanol, Glycerol 1,3-dichlorohydrin, Glycerol α,γ-dichlorohydrin, NSC 70982, Propylene dichlorohydrin, sym-Dichloroisopropyl alcohol, sym-Glycerol dichlorohydrin, α,γ-Dichlorohydrin, α-Dichlorohydrin
IUPAC Name 1,3-dichloropropan-2-ol
SMILES OC(CCl)CCl

Specifications

Quantity 50 g
InChI Formula InChI=1S/C3H6Cl2O/c4-1-3(6)2-5/h3,6H,1-2H2
Formula Weight 127.9796
Chemical Name or Material 1,3-Dichloropropan-2-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.