missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-2-methyl-5-nitrobenzene, 95%, Thermo Scientific™

Product Code. 10692063
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10692063

Brand: Thermo Scientific Maybridge KM08058EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 7149-69-1
Molecular Formula C7H5Cl2NO2
Molecular Weight (g/mol) 206.022
MDL Number MFCD00174267
InChI Key RUVCGWLHTVGNGI-UHFFFAOYSA-N
Synonym 2,6-dichloro-4-nitrotoluene, 1,3-dichloro-2-methyl-5-nitro-benzene, benzene, 1,3-dichloro-2-methyl-5-nitro, pubchem9506, maybridge1_005049, ksc496m1j
PubChem CID 251670
IUPAC Name 1,3-dichloro-2-methyl-5-nitrobenzene
SMILES CC1=C(C=C(C=C1Cl)[N+](=O)[O-])Cl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 206.03
Percent Purity 95%
Chemical Name or Material 1, 3-Dichloro-2-methyl-5-nitrobenzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.