missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-2-iodobenzene, 98%

Product Code. 15431588
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15431588

Brand: Thermo Scientific Alfa Aesar H55365.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19230-28-5 | C6H3Cl2I | 272.89 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19230-28-5
Molecular Formula C6H3Cl2I
Molecular Weight (g/mol) 272.89
MDL Number MFCD00060658
InChI Key ZMPGXSFTXBOKFM-UHFFFAOYSA-N
PubChem CID 140498
IUPAC Name 1,3-dichloro-2-iodobenzene
SMILES ClC1=CC=CC(Cl)=C1I

Specifications

Melting Point 64°C to 67°C
Quantity 25 g
Sensitivity Light sensitive
Synonym 2,6-dichloroiodobenzene, benzene, 1,3-dichloro-2-iodo, pubchem9581, acmc-1c3f6, ksc494i7b, 1,3-dichloro-2-iodo-benzene, #, benzene,1,3-dichloro-2-iodo, attercop-chm at115975
Formula Weight 272.9
Percent Purity 98%
Chemical Name or Material 1,3-Dichloro-2-iodobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.