missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-2-iodo-5-(trifluoromethyl)benzene, 97%, Thermo Scientific™

Product Code. 10180792
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
This item is not returnable. View return policy

Product Code. 10180792

Brand: Thermo Scientific Maybridge SEW01636DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175205-56-8
Molecular Formula C7H2Cl2F3I
Molecular Weight (g/mol) 340.89
MDL Number MFCD00084997
InChI Key RLCCFAVBZGICHD-UHFFFAOYSA-N
Synonym 1,3-dichloro-2-iodo-5-trifluoromethyl benzene, 3,5-dichloro-4-iodobenzotrifluoride, 3,5-dichloro-4-iodobenztrifluoride, buttpark 96\50-28, benzene,1,3-dichloro-2-iodo-5-trifluoromethyl, 1,3-bis chloranyl-2-iodanyl-5-trifluoromethyl benzene, 3,5-dichloro-4-iodo-alpha,alpha,alpha-trifluorotoluene
PubChem CID 2736838
IUPAC Name 1,3-dichloro-2-iodo-5-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC(Cl)=C(I)C(Cl)=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 340.89
Percent Purity 97%
Chemical Name or Material 1, 3-Dichloro-2-iodo-5-(trifluoromethyl)benzene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.