missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-2-iodo-5-(trifluoromethyl)benzene, 97%, Thermo Scientific™

Product Code. 10140842
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10140842 10 g 10g
10180792 1 g 1g
10747495 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10140842 Supplier Thermo Scientific Maybridge Supplier No. SEW01636EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175205-56-8
Molecular Formula C7H2Cl2F3I
Molecular Weight (g/mol) 340.89
MDL Number MFCD00084997
InChI Key RLCCFAVBZGICHD-UHFFFAOYSA-N
Synonym 1,3-dichloro-2-iodo-5-trifluoromethyl benzene, 3,5-dichloro-4-iodobenzotrifluoride, 3,5-dichloro-4-iodobenztrifluoride, buttpark 96\50-28, benzene,1,3-dichloro-2-iodo-5-trifluoromethyl, 1,3-bis chloranyl-2-iodanyl-5-trifluoromethyl benzene, 3,5-dichloro-4-iodo-alpha,alpha,alpha-trifluorotoluene
PubChem CID 2736838
IUPAC Name 1,3-dichloro-2-iodo-5-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC(Cl)=C(I)C(Cl)=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 340.89
Percent Purity 97%
Chemical Name or Material 1, 3-Dichloro-2-iodo-5-(trifluoromethyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.