missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-2-butene, Mixture of cis and trans Isomers 96%, Thermo Scientific™

Product Code. 10598312 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10598312 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10598312 Supplier Thermo Scientific Acros Supplier No. 406780050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 926-57-8
CAS Min % 95.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.16g/mL
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Molecular Formula C4H6Cl2
Linear Formula ClCH2CHCHClCH3
Refractive Index 1.4690 to 1.4730 (20°C, 589nm)
MDL Number MFCD00000861
Quantity 5 g
Fieser 01,214; 02,111; 10,134
Synonym 1,3-dichlorobutylene, unii-pwg1e5mrlj, 1,3-dichloro-2-butene, trans-1,3-dichlorobutene-2, 1,3-dichlorobut-2-ene, pwg1e5mrlj, 2-butene, 1,3-dichloro-, e, 2-butene, 1,3-dichloro, e-1,3-dichloro-2-butene, 1,3-dichlorobutene-2
InChI Key WLIADPFXSACYLS-DUXPYHPUSA-N
SMILES C\C(Cl)=C/CCl
IUPAC Name (2E)-1,3-dichlorobut-2-ene
Molecular Weight (g/mol) 124.99
PubChem CID 643911
Formula Weight 125
Percent Purity ≥95% (sum of cis and trans isomers)
Physical Form Liquid
Chemical Name or Material 1, 3-Dichloro-2-butene isomers, 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.