missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane, 97%

Product Code. 11327616
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11327616 25 g 25g
11307616 1 g 1g
11317616 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11327616 Supplier Thermo Scientific Alfa Aesar Supplier No. L11171.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 69304-37-6 | C12H28Cl2OSi2 | 315.425 g/mol

1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane is a silylating reagent used for simultaneous protection of 3'- and 5'-hydroxy functions of ribonucleosides. It is used in the analysis of Watson-Crick and Hoogstein base pairing in nucleotides. It is also employed in the formation of ribavirin chemical delivery systems. Further, it serves as a protecting group reagent for open-chain polyhydroxy compounds. In addition to this, it is involved in the preparation of cyclic bridged peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane is a silylating reagent used for simultaneous protection of 3′- and 5′-hydroxy functions of ribonucleosides. It is used in the analysis of Watson-Crick and Hoogstein base pairing in nucleotides. It is also employed in the formation of ribavirin chemical delivery systems. Further, it serves as a protecting group reagent for open-chain polyhydroxy compounds. In addition to this, it is involved in the preparation of cyclic bridged peptides.

Solubility
Miscible with dimethylformamide, hexane, ethyl acetate, chloroform and dichloromethane.

Notes
Incompatible with strong oxidizing agents and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69304-37-6
Molecular Formula C12H28Cl2OSi2
Molecular Weight (g/mol) 315.425
MDL Number MFCD00009655
InChI Key DDYAZDRFUVZBMM-UHFFFAOYSA-N
Synonym 1,3-dichloro-1,1,3,3-tetraisopropyldisiloxane, 1,3-dichlorotetraisopropyldisiloxane, 1,1,3,3-tetraisopropyl-1,3-dichlorodisiloxane, 1,3-dichloro-1,1,3,3-tetraisopropyl-disiloxane, disiloxane, 1,3-dichloro-1,1,3,3-tetrakis 1-methylethyl, 1,1,3,3-tetraisopropyl-1,3-dichlorosiloxane, chloro chlorodiisopropylsilyl oxy diisopropylsilane, chloro-chloro-di propan-2-yl silyl oxy-di propan-2-yl silane, tipdsicl2, tipdsi-cl2
PubChem CID 172404
IUPAC Name chloro-[chloro-di(propan-2-yl)silyl]oxy-di(propan-2-yl)silane
SMILES CC(C)[Si](C(C)C)(O[Si](C(C)C)(C(C)C)Cl)Cl

Specifications

Boiling Point 69°C to 70°C (0.5mmHg)
Quantity 25 g
Flash Point >110°C (230°F)
Refractive Index 1.454
Sensitivity Moisture sensitive
Density 0.986
UN Number UN1760
Beilstein 1934216
Solubility Information Miscible with dimethylformamide,hexane,ethyl acetate,chloroform and dichloromethane.
Formula Weight 315.43
Percent Purity 97%
Chemical Name or Material 1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane
Show More Show Less
compliance-icons
Product Identifier
  • 1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.