missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane, 97%

Product Code. 11317616
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11317616

Brand: Thermo Scientific Alfa Aesar L11171.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69304-37-6 | C12H28Cl2OSi2 | 315.425 g/mol

1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane is a silylating reagent used for simultaneous protection of 3'- and 5'-hydroxy functions of ribonucleosides. It is used in the analysis of Watson-Crick and Hoogstein base pairing in nucleotides. It is also employed in the formation of ribavirin chemical delivery systems. Further, it serves as a protecting group reagent for open-chain polyhydroxy compounds. In addition to this, it is involved in the preparation of cyclic bridged peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane is a silylating reagent used for simultaneous protection of 3′- and 5′-hydroxy functions of ribonucleosides. It is used in the analysis of Watson-Crick and Hoogstein base pairing in nucleotides. It is also employed in the formation of ribavirin chemical delivery systems. Further, it serves as a protecting group reagent for open-chain polyhydroxy compounds. In addition to this, it is involved in the preparation of cyclic bridged peptides.

Solubility
Miscible with dimethylformamide, hexane, ethyl acetate, chloroform and dichloromethane.

Notes
Incompatible with strong oxidizing agents and water.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69304-37-6
Molecular Formula C12H28Cl2OSi2
Molecular Weight (g/mol) 315.425
MDL Number MFCD00009655
InChI Key DDYAZDRFUVZBMM-UHFFFAOYSA-N
Synonym 1,3-dichloro-1,1,3,3-tetraisopropyldisiloxane, 1,3-dichlorotetraisopropyldisiloxane, 1,1,3,3-tetraisopropyl-1,3-dichlorodisiloxane, 1,3-dichloro-1,1,3,3-tetraisopropyl-disiloxane, disiloxane, 1,3-dichloro-1,1,3,3-tetrakis 1-methylethyl, 1,1,3,3-tetraisopropyl-1,3-dichlorosiloxane, chloro chlorodiisopropylsilyl oxy diisopropylsilane, chloro-chloro-di propan-2-yl silyl oxy-di propan-2-yl silane, tipdsicl2, tipdsi-cl2
PubChem CID 172404
IUPAC Name chloro-[chloro-di(propan-2-yl)silyl]oxy-di(propan-2-yl)silane
SMILES CC(C)[Si](C(C)C)(O[Si](C(C)C)(C(C)C)Cl)Cl

Specifications

Boiling Point 69°C to 70°C (0.5mmHg)
Quantity 5 g
UN Number UN1760
Beilstein 1934216
Flash Point >110°C (230°F)
Solubility Information Miscible with dimethylformamide,hexane,ethyl acetate,chloroform and dichloromethane.
Formula Weight 315.43
Percent Purity 97%
Refractive Index 1.454
Sensitivity Moisture sensitive
Density 0.986
Chemical Name or Material 1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.