missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromobutane, 98%

Product Code. 11349829
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349829 100 g 100g
11339829 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349829 Supplier Thermo Scientific Alfa Aesar Supplier No. B24123.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-80-2 | C4H8Br2 | 215.916 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-80-2
Molecular Formula C4H8Br2
Molecular Weight (g/mol) 215.916
MDL Number MFCD00000152
InChI Key XZNGUVQDFJHPLU-UHFFFAOYSA-N
Synonym butane, 1,3-dibromo, 1,3-butylene bromide, 1,3 dibromobutane, 1,3-dibrombutan, butane,1,3-dibromo, 1,3-dibromo-butan, 1,3-butylenebromide, 1,3-dibromo butane, 1,3-dibromo-butane, pubchem3866
PubChem CID 7889
IUPAC Name 1,3-dibromobutane
SMILES CC(CCBr)Br

Specifications

Density 1.789
Boiling Point 173°C to 176°C
Flash Point None
Refractive Index 1.5095
Quantity 100 g
Beilstein 1731231
Formula Weight 215.93
Percent Purity 98%
Chemical Name or Material 1,3-Dibromobutane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.