missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromobenzene, 97+%, Thermo Scientific Chemicals

Catalog Number 11440663
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
108-36-1
C6H4Br2
235.91
MFCD00000078
JSRLURSZEMLAFO-UHFFFAOYSA-N
m-dibromobenzene, benzene, 1,3-dibromo, benzene, m-dibromo, unii-74ef6kh8tc, 1,3-dibromo-benzene, 74ef6kh8tc, 1,3-dibromo benzene, 3,5-dibromobenzene, 1,3-dibrormobenzene, 1, 3-dibromobenzene
7927
CHEBI:37151
1,3-dibromobenzene
BrC1=CC(Br)=CC=C1
This item is not returnable. View return policy
108-36-1
C6H4Br2
235.91
MFCD00000078
JSRLURSZEMLAFO-UHFFFAOYSA-N
m-dibromobenzene, benzene, 1,3-dibromo, benzene, m-dibromo, unii-74ef6kh8tc, 1,3-dibromo-benzene, 74ef6kh8tc, 1,3-dibromo benzene, 3,5-dibromobenzene, 1,3-dibrormobenzene, 1, 3-dibromobenzene
7927
CHEBI:37151
1,3-dibromobenzene
BrC1=CC(Br)=CC=C1

1,3-Dibromobenzene is used in Suzuki coupling reaction for the preparation of [n]metacyclophanes. It is used to prepare 5-(3-bromo-phenyl)-2,4-di-tert-butoxy-pyrimidine using Pd(0) (PPh3)4 as a reagent. Further, it undergoes solid-supported potassium fluoride-aluminum oxide palladium-catalyzed polyarylation reaction with phenyl boronic acid to get conjugated polyaryls.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,3-Dibromobenzene is used in Suzuki coupling reaction for the preparation of [n]metacyclophanes. It is used to prepare 5-(3-bromo-phenyl)-2,4-di-tert-butoxy-pyrimidine using Pd(0) (PPh3)4 as a reagent. Further, it undergoes solid-supported potassium fluoride-aluminum oxide palladium-catalyzed polyarylation reaction with phenyl boronic acid to get conjugated polyaryls.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point °C to 7°C
Density 1.955
Boiling Point 219°C to 220°C
Flash Point 93°C (199°F)
Assay Percent Range 97%
Refractive Index 1.608
Quantity 25 g
UN Number UN3082
Beilstein 1904538
Solubility Information Immiscible with water.
Formula Weight 235.91
Percent Purity ≥97%
Physical Form Liquid
Chemical Name or Material 1, 3-Dibromobenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'