missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-2,2-dimethoxypropane, 99%, Thermo Scientific™

Product Code. 10436242 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10436242 1 g Glass bottle 1g
10570902 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10436242 Supplier Thermo Scientific Acros Supplier No. 355090010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22094-18-4
Molecular Formula C5H10Br2O2
Molecular Weight (g/mol) 261.94
MDL Number MFCD00051792
InChI Key CPAHOXOBYHMHDT-UHFFFAOYSA-N
Synonym 1,3-dibromoacetone dimethyl acetal, 1,3-acetone dibromide dimethyl acetal, 1,3-dibromo-2,2-dimethoxy-propane, 2-bromo-1-bromomethyl-1,1-dimethoxyethane, 1,3-dibromoacetone dimethyl ketal, propane, 1,3-dibromo-2,2-dimethoxy, pubchem2043, ag120 intermediates, acmc-1ck33, timtec-bb sbb039320
PubChem CID 2734196
IUPAC Name 1,3-dibromo-2,2-dimethoxypropane
SMILES COC(CBr)(CBr)OC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 65°C to 68°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 261.94
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 1, 3-Dibromo-2, 2-dimethoxypropane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.