missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dibromo-2,2-dimethoxypropane, 98%

Product Code. 11403544
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403544 25 g 25g
11493534 5 g 5g
11413544 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11403544 Supplier Thermo Scientific Alfa Aesar Supplier No. A11217.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22094-18-4 | C5H10Br2O2 | 261.941 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22094-18-4
Molecular Formula C5H10Br2O2
Molecular Weight (g/mol) 261.941
MDL Number MFCD00051792
InChI Key CPAHOXOBYHMHDT-UHFFFAOYSA-N
Synonym 1,3-dibromoacetone dimethyl acetal, 1,3-acetone dibromide dimethyl acetal, 1,3-dibromo-2,2-dimethoxy-propane, 2-bromo-1-bromomethyl-1,1-dimethoxyethane, 1,3-dibromoacetone dimethyl ketal, propane, 1,3-dibromo-2,2-dimethoxy, pubchem2043, ag120 intermediates, acmc-1ck33, timtec-bb sbb039320
PubChem CID 2734196
IUPAC Name 1,3-dibromo-2,2-dimethoxypropane
SMILES COC(CBr)(CBr)OC

Specifications

Melting Point 64°C to 67°C
Quantity 25 g
Beilstein 1698992
Formula Weight 261.95
Percent Purity 99%
Chemical Name or Material 1,3-Dibromo-2,2-dimethoxypropane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.